Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Round Bottom, Four-Neck Flasks with Thermometer Joint, Wilmad-LabGlass

Supplier: WILMAD/LABGLASS
LG-7360-162 LG-7360-130 LG-7360-164 LG-7360-186 LG-7360-110 LG-7360-100 LG-7360-134 LG-7360-114 LG-7360-136 LG-7360-103 LG-7360-180 LG-7360-171 LG-7360-182 LG-7360-172 LG-7360-104 LG-7360-116 LG-7360-106 LG-7360-118 LG-7360-109
These round bottom, borosilicate glass flasks have a center neck, two side necks 180° apart, and a third side neck 90° from the other two.

This third neck has a Standard Taper 10/30 joint to take a thermometer, gas inlet, or bleed tube. All four necks are vertical.



 

ORDER

Expand All / Collapse All
Capacity Center Neck [ST] Joint Side Neck [ST] Joints Supplier No. VWR Catalog Number Unit Price Quantity
500 mL 29/26 29/26 LG-7360-103 14204-262 Each Retrieving
500 mL 34/45 24/40 LG-7360-104 14208-402 Each Retrieving
1000 mL 24/40 24/40 LG-7360-106 14203-468 Each Retrieving
1000 mL 29/26 29/26 LG-7360-109 14207-232 Each Retrieving
1000 mL 34/45 24/40 LG-7360-110 14204-134 Each Retrieving
2000 mL 24/40 24/40 LG-7360-114 14206-610 Each Retrieving
2000 mL 29/42 24/40 LG-7360-116 14204-662 Each Retrieving
2000 mL 34/45 24/40 LG-7360-118 14203-638 Each Retrieving
5000 mL 24/40 24/40 LG-7360-130 14208-008 Each Retrieving
5000 mL 34/45 24/40 LG-7360-134 14204-622 Each Retrieving
5000 mL 45/50 24/40 LG-7360-136 14204-180 Each Retrieving
500 mL 34/45 29/42 LG-7360-162 14208-344 Each Retrieving
1000 mL 29/42 29/42 LG-7360-164 14203-896 Each Retrieving
2000 mL 29/26 29/26 LG-7360-171 14207-258 Each Retrieving
2000 mL 34/45 29/42 LG-7360-172 14208-412 Each Retrieving
3000 mL 45/50 29/42 LG-7360-180 14207-764 Each Retrieving
5000 mL 29/42 29/42 LG-7360-182 14208-964 Each Retrieving
5000 mL 45/50 29/42 LG-7360-186 14205-038 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping