Boron Standard for ICP, Certipur®, MilliporeSigma
Supplier: MilliporeSigma
Certipur®
1.70307.0100 1.70307.0100
EM1.70307.0100EA 116.59 USD
EM1.70307.0100 CA1.70307.0100
Boron Standard for ICP, Certipur®, MilliporeSigma
Standards Cationic Standards Boron Standards
H₃BO₃ in water
Certifications: Traceable to SRM from NIST. The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.