SHOWA 728R PVC, Chemical Resistant, 18" Gauntlet Glove, Showa
Supplier: Showa
728R-10
61200-223CS 822.72 USD
61200-223
SHOWA 728R PVC, Chemical Resistant, 18" Gauntlet Glove, Showa
Gloves
Tough PVC coating is designed for a wide variety of chemical, maintenance, and construction applications.
  • PVC, fully coated
  • Cotton interlocked knit lined
  • Rough grip
These gloves are designed for tough, abrasive applications, and for handling chemicals.
Certifications: ANSI Abrasion Level 4.