Cloxacillin sodium salt monohydrate

0215070225
75875-546EA 873.65 USD
75875-546
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate

Cloxacillin is a semi-synthetic antibiotic that is a chlorinated derivative of oxacillin. β-lactamase-resistant antibiotic.


Cloxacillin inhibits the last stage of bacterial cell wall synthesis by binding to certain penicillin-binding proteins (PBPs), which results in cell lysis. Cell lysis is mediated by bacterial cell wall autolytic enzymes.


Cloxacillin is used to treat infections caused by penicillin G-resistant staphylococci. It has been used in studies of topical antibacterial agent for burn and crush wounds. It is used as an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.

Formula: C₁₉H₁₈ClN₃O₅S
MDL Number: MFCD00150735
CAS Number: 7081-44-9
EINECS: 211-390-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR