J.T.Baker® HYDRA-POINT™, Volumetric Reagents for Water Determination by Karl Fischer Titration
Supplier: Avantor
BAKR6274-02 BAKR6274-68 BAKR6271-68 BAKR6271-02 BAKR6272-02 BAKR6275-68 BAKR6272-02 BAKR6272-68 BAKR6272-68 BAKR6275-68 BAKR6275-02 BAKR6274-02 BAKR6275-02 BAKR6271-02 BAKR6271-68 BAKR6274-68
JT6274-68CS 2266.35 USD
JT6274-68 CAJT6275-68 JT6272-68 CAJT6274-68 JT6272-2 JT6271-68 JT6275-68 JT6271-2 JT6274-2 JT6275-2 CAJT6271-2 CAJT6272-68 CAJT6275-2 CAJT6271-68 CAJT6274-2 CAJT6272-2
J.T.Baker® HYDRA-POINT™, Volumetric Reagents for Water Determination by Karl Fischer Titration
Karl Fischer Titrants and Reagents
No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free.
  • Pyridine-free
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titers that are more stable with less drift, helping you save time and reduce reagent usage
They are pyridine-free and specially formulated for existing application.