Mastic gum 5%, in alcohol

Supplier: Electron Microscopy Sciences

16591
101410-678EA 192.09 USD
101410-678
Mastic gum 5%, in alcohol
Mastic gum

Gum Mastic, Electron Microscopy Sciences, is available in concentrailns of 2.5%, 5%, or 10% in alcohol or 10% in isopropanol.

MDL Number: MFCD00131252
CAS Number: 61789-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR