BD BBL™ CDC Anaerobe 5% Sheep Blood Agar with Kanamycin and Vancomycin Prepared Media Stacker™ Plates, BD Diagnostics
Supplier: BD Diagnostics
221736
90006-459CS 233.62 USD
90006-459
BD BBL™ CDC Anaerobe 5% Sheep Blood Agar with Kanamycin and Vancomycin Prepared Media Stacker™ Plates, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Prepared plated media are designed for the isolation of microorganisms from samples or specimens and come in 100 x 10 mm or 100 x 15 mm petri-style configurations.
  • Plates contain a single medium
  • Interlocking, stackable design to minimize sliding or sample disruption
  • Ready for immediate use
Store at 2 to 8 °C (36 to 46 °F).