Merit™ Jeweler Style Forceps, Sklar
Supplier: Sklar
98-817
75996-348EA 32.21 USD
75996-348
Merit™ Jeweler Style Forceps, Sklar
Forceps
Merit™ Jeweler Style Forceps are grasping forceps used in microsurgical procedures.
  • Use for handling fine tissues
  • Multipurpose
  • Reusable
  • Nonsterile
The scissors have fine tips that can handle extremely fine tissues and materials that standard instruments may damage
Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee. Merit™ instruments deliver an economical solution for physicians and practices that require quality, durable instruments.
They are multipurpose thumb forceps with flat handles. This product is straight with smooth jaws and fine, pointed tips. It has a length of 4-1/4 inches. It is also known as the #5 style jeweler forceps.