Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SHOWA 3005 Powder-Free Medical Exam Nitrile Glove, 4 mil, SHOWA Best Glove

Supplier: Showa-Best Glove
3005PFL 3005PFM 3005PFS
Medical exam quality glove.

  • Class I medical exam
  • Powder-free, 4mil Nitrile construction
  • 9.5"" length

Class I Medical glove for patient examinations, handling blood borne pathogens, laboratory work, and use in critical environments.

Certifications: In Compliance 21CFR, parts 170-199.



 

ORDER

Expand All / Collapse All
Glove Size Length Thickness Glove Color Supplier No. VWR Catalog Number Unit Price Quantity
Small 30.5 cm (12") 4 mil Blue 3005PFS 10847-450 Box of 100 Retrieving
Case of 1000 Retrieving
Medium 30.5 cm (12") 4 mil Blue 3005PFM 10847-452 Box of 100 Retrieving
Case of 1000 Retrieving
Large 30.5 cm (12") 4 mil Blue 3005PFL 10847-454 Box of 100 Retrieving
Case of 1000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/Hand-specific Ambidextrous
Disposable/Reusable Disposable
Latex/Latex-free Latex-Free
Powdered/Powder-free Powder-Free
Finish Textured
Food Contact Approved +
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping