Butyl-4-hydroxybenzoate NF

Supplier: ENCORE SCIENTIFIC MS

Synonyms: 4-Hydroxybenzoic acid butyl ester Butylparaben Butyl parahydroxybenzoate

BU005-00112000 BU005-00125000 BU005-00112000 BU005-00125000
103566-010EA 1192 USD InStock
103566-010 76027-890 76027-888 103566-012
Butyl-4-hydroxybenzoate NF
Butyl 4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 156 to 157°C (3,5 torr)
Melting Pt: 69-71°C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 183,60°C
Storage Temperature: Ambient
MDL Number: MFCD00016478
CAS Number: 94-26-8
EINECS: 202-318-7
UN: 0000
Merck Index: 13,01583

Order Now

Specification Test Results

Appearance White Crystalline Powder
Assay 99%+
Idenitification by FTIR Conforms
Total Impurities 1.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR