L(+)-Arginine hydrochloride USP

Supplier: ENCORE SCIENTIFIC MS

Synonyms: (S)-(+)-Arginine hydrochloride H-L-Arg-OH.HCl H-Arg-OH.HCl

AR002-00112000 AR002-00105000 AR002-00112000 AR002-00105000
103565-976EA 1116 USD InStock
103565-976 103565-974 76027-836 76027-858
L(+)-Arginine hydrochloride USP
L(+)-Arginine hydrochloride
Formula: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Boiling Pt: 409,1°C (760 mmHg)
Melting Pt: 235°C
Density: 1.42 g/cm³ (20 °C)
Flash Pt: 201,2°C
Storage Temperature: Ambient
MDL Number: MFCD00064550
CAS Number: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785

Order Now

Specification Test Results

Appearance White crystals or white crystalline powder
Assay 0.9992
Identification Per USP Conforms
pH 5.25
Specific Optical Rotation +22.76°
Loss on Drying 0.0008
Residue on Ignition 0.1%
Heavy Metals 0.0005%
Chloride 0.169

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR