Sorbic acid, resin FCC

Supplier: ENCORE SCIENTIFIC MS

Synonyms: 2,4-Hexadienoic acid(E,E)-2,4-Hexadienoic acid(2E,4E)-2,4-Hexadienoic acidtrans,trans-2,4-Hexadienoic acid

SO033-05212000 SO033-05212000 SO033-05200500 SO033-05200500 SO033-05202500 SO033-05202500
76028-188EA 1181.13 USD InStock
76028-188 103566-356 76028-184 103566-352 76028-186 103566-354
Sorbic acid, resin FCC
Sorbic acid
Formula: CH₃CH=CHCH=CHCOOH
MW: 112,13 g/mol
Boiling Pt: 233°C (760 mmHg)
Melting Pt: 132-135°C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 139.9°C
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://us.vwr.com/stibo/web/std.lang.all/29/27/2302927.jpg

Order Now

Specification Test Results

Assay 0.998
Appearance White, crystalline powder
Identification by UV Pass
Identification by FTIR Conforms

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR