Potassium sorbate, microcrystalline NF

Supplier: ENCORE SCIENTIFIC MS

Synonyms: (2E,4E)-2,4-Hexadienoic acid potassium salt (E,E)-2,4-Hexadienoic acid potassium salt 2,4-Hexadienoic acid potassium salt Sorbic acid potassium salt trans,trans-2,4-Hexadienoic acid potassium salt Potassium (E,E)-hexa-2,4-dienoate Potassium (2E,4E)-hexa-2,4-dienoate Potassium trans,trans-hexa-2,4-dienoate

PO010-03112000 PO010-03100500 PO010-03102500 PO010-03100500 PO010-03102500 PO010-03112000
76028-114EA 1002.96 USD InStock
76028-114 76028-110 103566-060 103566-284 76028-112 103566-062
Potassium sorbate, microcrystalline NF
Potassium sorbate

Antimicrobial preservative used in many food and personal care products. Potassium sorbate also stabilizes wine at the end of the fermentation process.

Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 270°C
Density: 1.36 g/cm3
Storage Temperature: Ambient
CAS Number: 24634-61-5
EINECS: 246-376-1
Merck Index: 13,07756
https://us.vwr.com/stibo/web/std.lang.all/71/23/2297123.jpg

Order Now

Specification Test Results

Appearance White rod-shaped pellets
Assay Min. 98.0%
Loss on Drying Max. 1.0%
Acidity (as sorbic acid) Max. 1.0%
Alkalinity (as K2CO3) Max. 1.0%
Lead Max 0.0002%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR