You Searched For: Wright's stain


17  results were found

SearchResultCount:"17"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR International
Description: VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.
Catalog Number: (10143-062)
Supplier: VWR International
Description: VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.

Supplier: VWR International
Description: VWR® brand hematology stain offerings satify the needs of laboratory professionals following manual, rapid stain or automated protocols.
Catalog Number: (34172-114)
Supplier: MilliporeSigma
Description: For differential blood staining.

Supplier: MilliporeSigma
Description: HARLECO® Polychromic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation.
Catalog Number: (EM1.09278.0025)
Supplier: MilliporeSigma
Description: WRight's eosine methylene blue for microscopy, Glass bottle, 25g, The dry mixture of the Wright's eosin methylene blue dye is used for the self-preparation of the Wright's staining solution

Catalog Number: (34172-112)
Supplier: MilliporeSigma
Description: HARLECO® Designed for use with the Ames Hema-Tek® Stainer

Supplier: Thermo Scientific Chemicals
Description: WRIGHT STAIN PURE CERTIFIED 100G

Supplier: TCI America
Description: [for Microscopy and Bacteriology] CAS Number: 68988-92-1 MDL Number: MFCD00082143 Form: Crystal Color: Deep Green Flash Point (°C): 11
Supplier: Epredia
Description: Utilize Epredia™ Wright Stain for peripheral blood and bone marrow smears.

SDS

Supplier: Thermo Scientific Chemicals
Description: Useful stain for blood and for bone marrow films
Supplier: Azer Scientific Inc.
Description: Wright stain is a polychromatic hematology stain consisting of eosin and methylene blue.

SDS Product available on GSA Advantage® Minority or Woman-Owned Business Enterprise

Supplier: Ricca Chemical
Description: Regular. Container: Glass.
Supplier: Electron Microscopy Sciences
Description: For use as a blood stain

SDS Minority or Woman-Owned Business Enterprise

Supplier: Ricca Chemical
Description: Rapid. Container: Glass.
Catalog Number: (100504-340)
Supplier: Electron Microscopy Sciences
Description: For differentiation of blood corpuscles

Minority or Woman-Owned Business Enterprise


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 17
no targeter for Bottom