Uranyl acetate 4% in aqueous solution Ready-to-Use
Supplier: Electron Microscopy Sciences
|
Warning
|
22400-4
102092-288EA
144.37
USD
102092-288
Uranyl acetate 4% in aqueous solution Ready-to-Use
Uranyl acetate
Pre-made and ready to use uranyl acetate solution are avaialable with concentrations of 1%, 2%, 3%, or 4%.
Other concentrations are available upon request.
Formula:
C₄H₈O₆U Boiling Pt: 100 °C (212 °F) Storage Temperature: Ambient |
CAS Number:
541-09-3 UN: 2910 ADR: 7, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...