    Sort by:
  • Search Within Results
L-Methionine, [Methyl-3H]
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  MP Biomedicals
Description:   Sp. Act. 5-15 Ci/mmol; 185-555 GBq/mmol Purity 94% Ethanol: water solution (3:7). Containing 10 mM 2-mercaptoethanol Shipped in dry ice.