Anti-PRDX5 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-5755
102026-478EA 718.77 USD
102026-478
Anti-PRDX5 Rabbit Polyclonal Antibody
Antibodies
PRDX5 is a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in different tissues under normal conditions and during inflammatory processes. This protein interacts with peroxisome receptor 1. The crystal structure of this protein in its reduced form has been resolved to 1.5 angstrom resolution. This gene uses alternate in-frame translation initiation sites to generate mitochondrial or peroxisomal/cytoplasmic forms.

WB: 1 ug/ml

Type: Primary
Antigen: PRDX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR