Anti-PRDM15 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-1966
102018-900EA 718.77 USD
102018-900
Anti-PRDM15 Rabbit Polyclonal Antibody
Antibodies
PRDM15 may be involved in transcriptional regulation.

WB: 1 ug/ml

Type: Primary
Antigen: PRDM15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR