Anti-PRD Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-3199
102021-366EA 718.77 USD
102021-366
Anti-PRD Rabbit Polyclonal Antibody
Antibodies
Prd is a pair-rule protein expressed in a segmentally repeating pattern to define the polarity of embryonic segments.

WB: 0.5 ug/ml

Type: Primary
Antigen: PRD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR