Anti-pRB Mouse Monoclonal Antibody [clone: 2C4]

10R-P122A
101983-784EA 686.54 USD
101983-784
Anti-pRB Mouse Monoclonal Antibody [clone: 2C4]
Cyclin-dependent kinases (Cdks) play important roles in the regulation of the cell cycle. RB protein (pRB) is phosphorylated by cyclin D-Cdk4/Cdk6 and cyclin A/cyclin E-Cdk2 during the G1/S transition. This phosphorylation causes the inactivation of the growth inhibitory functions of pRB. pRB undergo phosphorylation and attendant functional inactivation, the cell proceed into late G1. Cyclin D-Cdk4 specifically phosphorylates Ser780 in pRB, while cyclin E-Cdk2 and cyclin A-Cdk2 does not.

WB: 1 ug/ml

Type: Primary
Antigen: pRB
Clonality: Monoclonal
Clone: 2C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR