Anti-PPP1R2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-10305
102016-394EA 718.77 USD
102016-394
Anti-PPP1R2 Rabbit Polyclonal Antibody
Antibodies
PPP1R2 is an inhibitor of protein-phosphatase 1.

Optimal conditions to be determined by the end user

Type: Primary
Antigen: PPP1R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR