Anti-PPME1 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-3708
102022-384EA 718.77 USD
102022-384
Anti-PPME1 Rabbit Polyclonal Antibody
Antibodies
Protein phosphatase methylesterase-1 catalyzes the demethylation of the protein phosphatase-2A catalytic subunit.

WB: 1 ug/ml

Type: Primary
Antigen: PPME1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR