Anti-PPID Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2388
102019-744EA 718.77 USD
102019-744
Anti-PPID Rabbit Polyclonal Antibody
Antibodies
PPID is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein has been shown to possess PPIase activity and, similar to other family members, can bind to the immunosuppressant cyclosporin A.

WB: 1 ug/ml

Type: Primary
Antigen: PPID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR