Anti-PPAPDC2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-6945
102028-858EA 498.46 USD
102028-858
Anti-PPAPDC2 Rabbit Polyclonal Antibody
Antibodies
PPAPDC2 is the phosphatase that dephosphorylates presqualene diphosphate (PSDP) into presqualene monophosphate (PSMP), suggesting that it may be indirectly involved in innate immunity. PSDP is a bioactive lipid that rapidly remodels to presqualene monophosphate PSMP upon cell activation. PPAPDC2 displays diphosphate phosphatase activity with a substrate preference for PSDP > FDP > phosphatidic acid.

WB: 1 ug/ml

Type: Primary
Antigen: PPAPDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR