Anti-PP2447 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-1269
102017-506EA 626.46 USD
102017-506
Anti-PP2447 Rabbit Polyclonal Antibody
Antibodies
The function of PP2447 protein is not widely studied, and is yet to be elucidated fully.

WB: 2.5 ug/ml

Type: Primary
Antigen: PP2447
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR