Anti-POU5F1 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-8464
102031-896EA 718.77 USD
102031-896
Anti-POU5F1 Rabbit Polyclonal Antibody
Antibodies
POU5F1 is a POU transcription factor expressed by early embryo cells and germ cells. It determines paracrine growth factor signaling from stem cells to the trophectoderm.

WB: 0.2-1 ug/ml

Type: Primary
Antigen: POU5F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR