Anti-Porcn Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-8827
102032-622EA 718.77 USD
102032-622
Anti-Porcn Rabbit Polyclonal Antibody
Antibodies
Porcn modulates the processing of Wnt proteins. Probable protein-cysteine N-palmitoyltransferase that palmitoylates Wnt family members.

Optimal conditions to be determined by the end user

Type: Primary
Antigen: Porcn
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR