Anti-POLR3F Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2038
102019-044EA 718.77 USD
102019-044
Anti-POLR3F Rabbit Polyclonal Antibody
Antibodies
POLR3F is one of more than a dozen subunits forming eukaryotic RNA polymerase III (RNA Pol III), which transcribes 5S ribosomal RNA and tRNA genes. This protein has been shown to bind both TFIIIB90 and TBP, two subunits of RNA polymerase III transcription initiation factor IIIB (TFIIIB). Unlike most of the other RNA Pol III subunits, the encoded protein is unique to this polymerase.

WB: 1 ug/ml

Type: Primary
Antigen: POLR3F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR