Anti-POLR3B Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2297
102019-562EA 718.77 USD
102019-562
Anti-POLR3B Rabbit Polyclonal Antibody
Antibodies
POLR3B belongs to the RNA polymerase beta chain family. DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. POLR3B is the second largest core component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It is proposed to contribute to the polymerase catalytic activity and forms the polymerase active center together with the largest subunit. Pol III is composed of mobile elements and RPC2 is part of the core element with the central large cleft and probably a clamp element that moves to open and close the cleft.

WB: 1 ug/ml

Type: Primary
Antigen: POLR3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR