Anti-POLR2H Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-1052
102016-866EA 626.46 USD
102016-866
Anti-POLR2H Rabbit Polyclonal Antibody
Antibodies
This gene encodes one of the essential subunits of RNA polymerase II that is shared by the other two eukaryotic DNA-directed RNA polymerases, I and III. This gene encodes a member of the E2F transcription factor protein family. E2F family members play a crucial role in control of the cell cycle and of the action of tumor suppressor proteins.

WB: 5 ug/ml

Type: Primary
Antigen: POLR2H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR