Anti-POLR2F Mouse Monoclonal Antibody [clone: 2780C3a]

Supplier: Fitzgerald Industries
10R-1369
101979-238EA 583.23 USD
101979-238
Anti-POLR2F Mouse Monoclonal Antibody [clone: 2780C3a]
Antibodies
This gene encodes the sixth largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes, that is also shared by the other two DNA-directed RNA polymerases. In yeast, this polymerase subunit, in combination with at least two other subunits, forms a structure that stabilizes the transcribing polymerase on the DNA template.

To be optimized by the end user.

Type: Primary
Antigen: POLR2F
Clonality: Monoclonal
Clone: 2780C3a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR