Anti-POLR1B Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2934
102020-836EA 718.77 USD
102020-836
Anti-POLR1B Rabbit Polyclonal Antibody
Antibodies
Eukaryotic RNA polymerase I (pol I) is responsible for the transcription of ribosomal RNA (rRNA) genes and production of rRNA, the primary component of ribosomes. Pol I is a multisubunit enzyme composed of 6 to 14 polypeptides, depending on the species.

WB: 1 ug/ml

Type: Primary
Antigen: POLR1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR