Anti-POLL Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-5637
102026-242EA 718.77 USD
102026-242
Anti-POLL Rabbit Polyclonal Antibody
Antibodies
POLL is a repair polymerase.It is involved in base excision repair (BER) responsible for repair of lesions that give rise to abasic (AP) sites in DNA. Has both DNA polymerase and terminal transferase activities. Has a 5'-deoxyribose-5-phosphate lyase (dRP lyase) activity.

WB: 1 ug/ml

Type: Primary
Antigen: POLL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR