Anti-POLE4 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-8343
102031-654EA 718.77 USD
102031-654
Anti-POLE4 Rabbit Polyclonal Antibody
Antibodies
POLE4 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.

WB: 0.2-1 ug/ml

Type: Primary
Antigen: POLE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR