Anti-POLDIP3 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-4847
102024-662EA 718.77 USD
102024-662
Anti-POLDIP3 Rabbit Polyclonal Antibody
Antibodies
POLDIP3 is a protein that interacts with the DNA polymerase delta p50 subunit. This protein is a specific target of S6 kinase 1 and regulates cell growth. Two transcript variants that encode different protein isoforms have been identified.

WB: 1 ug/ml

Type: Primary
Antigen: POLDIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR