Anti-POLDIP2 Mouse Monoclonal Antibody [clone: 2765C3a]

Supplier: Fitzgerald Industries
10R-1710
101979-920EA 583.23 USD
101979-920
Anti-POLDIP2 Mouse Monoclonal Antibody [clone: 2765C3a]
Antibodies
This gene encodes a protein that interacts with the DNA polymerase delta p50 subunit. The encoded protein also interacts with proliferating cell nuclear antigen. Some transcripts of this gene overlap in a tail-to-tail orientation with the gene for tumor necrosis factor, alpha-induced protein 1 (TNFAIP1).

To be optimized by the end user.

Type: Primary
Antigen: POLDIP2
Clonality: Monoclonal
Clone: 2765C3a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR