Anti-POLD2 Mouse Monoclonal Antibody [clone: 2762C3a]

Supplier: Fitzgerald Industries
10R-1422
101979-344EA 967.38 USD
101979-344
Anti-POLD2 Mouse Monoclonal Antibody [clone: 2762C3a]
Antibodies
the POLD2 gene encodes DNA polymerase delta subunit 2 enzyme . It is a component of the DNA polymerase delta complex.

To be optimized by the end user.

Type: Primary
Antigen: POLD2
Clonality: Monoclonal
Clone: 2762C3a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR