Anti-POLD2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-5521
102026-010EA 718.77 USD
102026-010
Anti-POLD2 Rabbit Polyclonal Antibody
Antibodies
This protein is a DNA polymerase delta complex, it is involved in DNA replication and repair.

WB: 1 ug/ml

Type: Primary
Antigen: POLD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR