Anti-POLA2 Mouse Monoclonal Antibody [clone: 2759C3a]

Supplier: Fitzgerald Industries
10R-1567
101979-634EA 583.23 USD
101979-634
Anti-POLA2 Mouse Monoclonal Antibody [clone: 2759C3a]
Antibodies
This gene may play an essential role at the early stage of chromosomal DNA replication by coupling the polymerase alpha/primase complex to the cellular replication machinery.

WB: 0.2-2 ug/ml, ICC: 2-100 ug/ml

Type: Primary
Antigen: POLA2
Clonality: Monoclonal
Clone: 2759C3a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR