Anti-POGZ Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
20R-1216
101985-794EA 626.46 USD
101985-794
Anti-POGZ Rabbit Polyclonal Antibody
Antibodies
POGZ appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid systemThe protein encoded by this gene appears to be a zinc finger protein containing a transposase domain at the C-terminus. This protein was found to interact with the transcription factor SP1 in a yeast two-hybrid system.

WB: 5 ug/ml

Type: Primary
Antigen: POGZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR