Anti-PODXL Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-1719
102018-406EA 626.46 USD
102018-406
Anti-PODXL Rabbit Polyclonal Antibody
Antibodies
PODXL is involved in the regulation of both adhesion and cell morphology and cancer progression. Function as an anti-adhesive molecule that maintains an open filtration pathway between neighboring foot processes in the podocyte by charge repulsion. Acts as a pro-adhesive molecule, enhancing the adherence of cells to immobilized ligands, increasing the rate of migration and cell- cell contacts in an integrin-dependent manner. Induces the formation of apical actin-dependent microvilli. Involved in the formation of a preapical plasma membrane subdomain to set up inital epithelial polarization and the apical lumen formation during renal tubulogenesis. Plays a role in cancer development and aggressiveness by inducing cell migration and invasion through its interaction with the actin-binding protein EZR. Affects EZR- dependent signaling events, leading to increased activities of the MAPK and PI3K pathways in cancer cells. Belongs to the podocalyxin family.

WB: 5 ug/ml

Type: Primary
Antigen: PODXL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR