Anti-PNRC2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-3985
102022-938EA 718.77 USD
102022-938
Anti-PNRC2 Rabbit Polyclonal Antibody
Antibodies
PNRC2 is involved in nonsense-mediated mRNA decay (NMD) by acting as a bridge between the mRNA decapping complex and the NMD machinery.PNRC2 may act by targeting the NMD machinery to the P-body and recruiting the decapping machinery to aberrant mRNAs. PNRC2 is required for UPF1/RENT1 localization to the P-body. PNRC2 also acts as a nuclear receptor coactivator. PNRC2 may play a role in controlling the energy balance between energy storage and energy expenditure.

WB: 1 ug/ml

Type: Primary
Antigen: PNRC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR