Anti-PNPLA8 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2261
102019-490EA 718.77 USD
102019-490
Anti-PNPLA8 Rabbit Polyclonal Antibody
Antibodies
Phospholipase A2 catalyzes cleavage of fatty acids from phospholipids, thereby regulating membrane physical properties and the release of lipid second messengers and growth factors.

WB: 1 ug/ml

Type: Primary
Antigen: PNPLA8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR