Anti-PNPLA5 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-2827
102020-622EA 718.77 USD
102020-622
Anti-PNPLA5 Rabbit Polyclonal Antibody
Antibodies
Human patatin-like phospholipases, such as PNPLA5, have been implicated in regulation of adipocyte differentiation and have been induced by metabolic stimuli.

WB: 1 ug/ml

Type: Primary
Antigen: PNPLA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR