Anti-PNN Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-6051
102027-070EA 718.77 USD
102027-070
Anti-PNN Rabbit Polyclonal Antibody
Antibodies
PNN is the transcriptional activator binding to the E-box 1 core sequence of the E-cadherin promoter gene; the core-binding sequence is 5'CAGGTG-3'. PNN is capable of reversing CTBP1-mediated transcription repression.

WB: 1 ug/ml

Type: Primary
Antigen: PNN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR