Anti-PNKP Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-4477
102023-922EA 718.77 USD
102023-922
Anti-PNKP Rabbit Polyclonal Antibody
Antibodies
This locus represents a gene involved in DNA repair. In response to ionizing radiation or oxidative damage, the protein encoded by this locus catalyzes 5' phosphorylation and 3' dephosphorylation of nucleic acids. Mutations at this locus have been associated with microcephaly, seizures, and developmental delay.

WB: 1 ug/ml

Type: Primary
Antigen: PNKP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR