Anti-PMN Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
20R-PR052
101987-336EA 619.08 USD
101987-336
Anti-PMN Rabbit Polyclonal Antibody
Antibodies
Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.

Type: Primary
Antigen: PMN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR