Anti-PMM1 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-3894
102022-756EA 718.77 USD
102022-756
Anti-PMM1 Rabbit Polyclonal Antibody
Antibodies
Phosphomannomutase catalyzes the conversion between D-mannose 6-phosphate and D-mannose 1-phosphate which is a substrate for GDP-mannose synthesis. GDP-mannose is used for synthesis of dolichol-phosphate-mannose, which is essential for N-linked glycosylat

WB: 1 ug/ml

Type: Primary
Antigen: PMM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR