Anti-PMCH Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
20R-1341
101986-044EA 626.46 USD
101986-044
Anti-PMCH Rabbit Polyclonal Antibody
Antibodies
MCH may act as a neurotransmitter or neuromodulator in a broad array of neuronal functions directed toward the regulation of goal-directed behavior, such as food intake, and general arousal. May also have a role in spermatocyte differentiation.

Optimal conditions to be determined by the end user

Type: Primary
Antigen: PMCH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR