Anti-PM20D2 Rabbit Polyclonal Antibody

Supplier: Fitzgerald Industries
70R-4116
102023-200EA 718.77 USD
102023-200
Anti-PM20D2 Rabbit Polyclonal Antibody
Antibodies
PM20D2 belongs to the peptidase M20A family. The function of PM20D2 remains unknown.

WB: 1 ug/ml

Type: Primary
Antigen: PM20D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR